Learn More
AMORSUI INC Bi Layered Face Mask 9.5" x 5"
Supplier:  AMORSUI INC AMS13
Encompass_Preferred
This fire resistant face mask is made of 6.4 oz Pique Dual Hazard Knit (45% Lyocell, 40% Modacrylic, 15% para-aramid) The dimensions for the size standard are 9.5" x 5". Other features of this product are machine washability, double-layered for extra protection, fast drying, lightweight, breathable, odor resistant, with superior moisture wicking. The manufacturer recommends using a knot to adjust the length of the ear loops for a tighter fit as needed.This product is certified fire resistant (NFPA 2112, EN61481-1-2, and ISO 11612), and provides arc protection (NFPA 70E) for the lifetime of the garment. CAT 2 = 9.5 CAL/CL2 ATPV
Catalog No. NC1827936
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.