Learn More
Frataxin Mouse anti-Human, Unconjugated, Clone: 4F9, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABN2313100UG
Description
Frataxin Monoclonal antibody specifically detects Frataxin in Human samples. It is validated for Western Blotting.Specifications
Frataxin | |
Monoclonal | |
Unconjugated | |
EC:1.16.3.1;Friedreich ataxia protein;Fxn | |
Mouse | |
∼14 and 18 kDa observed; 23.13 kDa calculated (full length). Uncharacterized bands may be observed in some lysate(s). | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
4F9 | |
NP_000135 | |
FXN, FRDA, X25 | |
Full-length human recombinant Frataxin. | |
Protein G Purified | |
Neuroscience | |
Clone 4F9 is a mouse monoclonal antibody that specifically detects mature and intermediate forms of human Frataxin. | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
IgG2b κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.