Learn More
MilliporeSigma FREUND'S COMPLETE ADJUVANT
Supplier:  MilliporeSigma AR001
Encompass
Composition: 85% Drakeol 6VR (Mineral Oil) 15% Mannide Monooleate (Arlacel A) 20 mg Mycobacterium 50 mL
Catalog No. 50-554-75
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.