Learn More
Electron Microscopy Sciences Friedman Bone Rongeur
Supplier:  Electron Microscopy Sciences 7798001
Encompass_Preferred
Mini Bone Rongeur, Curved. The jaw width is 2 mm and it is 5 1/2" long. Made from stainless steel with a polished finish. Mini bone rongeur, curved, the jaw width is 2mm.
OAL: 5.5"
Catalog No. 50-301-89
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.