Learn More
MilliporeSigma™ Supelco™ Frit for Omnifit™ Column Endpiece
Frit's pore diameter and surface structures should be optimized to avoid clogging and abrasion of chromatographic particles.
Supplier: MilliporeSigma™ Supelco™ 56271U
Description
- End pieces are equipped with porous frits
- Frits can be made of sintered metal, glass and plastic
- Designed to be easily exchangeable
- Can also be used to keep packing material in place
Specifications
Columns - 15 mm Bore | |
20 Pk. | |
25 μm |
Polyethylene | |
Frit |
Omnifit is a registered trademark of Diba Industries