Learn More
Electron Microscopy Sciences Formvar/Carbon Support Film 10 nm/20-30 nm thick on Gilder F1 Finder Nickel Grid
Supplier:  Electron Microscopy Sciences FCF200F1NISC
Encompass_Preferred
A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. Coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. NOTE: All film is laid on the shiny side of the grid. Formvar is approximately 10 nm thick with 20 to 30 nm of carbon.
Grid: Nickel
Catalog No. 50-190-3496
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.