Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Drawer Rack for 96 Deep-Well Plates or 1 3/4" Max H Boxes, 7 x 4 Array
Supplier: Cole-Parmer 0440083
These upright freezer drawer racks provide quick storage and retrieval of plates and microtube boxes. They are crafted of corrosion-resistent stainless steel for durablity.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.