Learn More
TARGETMOL CHEMICALS INC FSCPX 5MG
Supplier:  TARGETMOL CHEMICALS INC T39039
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. FSCPX is a potent and selective irreversible antagonist of the A1 adenosine receptor (A1AR) exhibiting low nanomolar binding potency. Additionally FSCPX can effectively decrease the interstitial adenosine level in the guinea pig atrium thereby modifying the effect of NBTI a nucleoside transport inhibitor. purity: 98%
Catalog No. 50-258-0072
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.