Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Full Finger Anti-Fatigue Thermo Gloves

Catalog No. p-7084606 Shop All Impacto Protective Products Products
Click to view available options
Size:
2X-Large
Large
Medium
Small
X-Large
X-Small
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves provide light but firm compression to counteract tissue swelling, providing temporary relief commonly associated with repetitive strain injuries. Designed to allow full ventilation during long-lasting, repetitive tasks.

  • Natural pain relief for prevention of injuries during work or play
  • Designed to allow full ventilation during repetitive tasks, for extended periods of time
  • Light but firm therapeutic compression counteracts tissue swelling to provide temporary relief from repetitive strain injuries (RSI)
  • Promotes optimal muscle function at 1°C higher than the body’s normal resting temperature
  • Ideal for repetitive jobs such as sewing, or for wearing inside other gloves when handling frozen products

Specifications

Color Black
Finger Style Full Finger
For Use With (Application) Temporary Pain Relief from Repetative Strain Injuries (RSI)
Glove Cuff Style Hook and Loop
Material Nylon
Series TS299
Size X-Small
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.