Learn More
Walter Products™ Full-size Disarticulated Human Skeleton with Muscles Model
High quality, life-size disarticulated skeleton model with color-coded spinal column.
Supplier: Walter Products™ B10247
This item is not returnable.
View return policy
Description
- All 206 bones of this full-size disarticulated skeleton are cast individually with high anatomical detail
- Skeleton has one hand and one foot that are articulated with wire and a 3-part skull with removable skull cap and spring-mounted mandible
- Left side of the skeleton is hand-painted to realistically display the muscle origins in red and muscle insertions in blue
- Spinal column is assembled, with the different regions color-coded and labeled
- Color poster of skeletal system included
- Skeleton is made exclusively from PVC
Specifications
Anatomical Model | |
PVC |