Learn More
Chemglass Life Sciences Funnel, Vial, PTFE, 1.9" OD, 22-400 GPI Thread
SDP
Supplier:  Chemglass Life Sciences CG176107
Encompass_Preferred
Funnel, Vial, PTFE, 1.9" OD, 22-400 GPI Thread. Funnel machined from PTFE screws securely onto any standard laboratory vial having a GPI thread. Design allows for easy, safe and mess free transfer of product.
Catalog No. 50-153-1233
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.