Learn More
Molecular Probes™ Fura-2 Calcium Imaging Calibration Kit (Zero to 10 mM CaEGTA, 50 μM Fura-2)

Specifications
Content And Storage
Store in refrigerator 2°C to 8°C and protect from light.
Detection Method
Fluorescence
For Use With (Application)
Calcium Imaging Calibration
For Use With (Equipment)
Fluorescence Microscope
Product Type
Reagent
Dye Type
Ion Calibration Reagent
Quantity
1 kit
Shipping Condition
Room Temperature
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.