Learn More
Apexbio Technology LLC Fusaric Acid,250mg CAS# 536-69-6
SDP
Supplier:  Apexbio Technology LLC C5062250
Encompass_Preferred
Fusaric acid is a mycotoxin produced by several species of Fusarium [1]. Mycotoxins are biologically active secondary fungal metabolites found as contaminants of food- and feedstuff. Mycotoxin is capable of causing disease and death in both humans and animals.Fusaric acid is a potent inhibitor of dopamine β-hydroxylase. Fusaric acid uncompetitively inhibited the activity of dopamine β-hydroxylase with an IC50 of 30 μM in an. Fusaric acid lowered endogenous levels of norepinephrine and epinephrine in brain, spleen, heart, and adrenal glands. Fusaric acid inhibited dopamine β-hydroxylase activity in adrenal medulla in vivo [2]. Fusaric acid altered brain and pineal neurotransmitters. In the brain and pineal gland of rats, intraperitoneally (100 mg/kg) administration of fusaric acid increased the level of 5HT, 5-hydroxyindoleacetic acid (5HIAA), tyrosine, and dopamine [3]. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0223
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.