Learn More
Research Products International Corp G-418 Disulfate 50 mg/ml Solution, 20 Milliliters
Aminoglycoside antibiotic used as a selective agent in transformation experiments. Dissolved in Molecular Biology Grade Water and sterile filtered.