Learn More
Research Products International Corp G-418 Disulfate 50 mg/ml Solution, 50 Milliliters
SDP
Supplier:  Research Products International Corp G6450050.0
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Aminoglycoside antibiotic used as a selective agent in transformation experiments. Dissolved in Molecular Biology Grade Water and sterile filtered.
Catalog No. 50-488-696
missing translation for 'QuantityLeft'