Learn More
Enzo Life Sciences Gabexate . mesylate (50mg). CAS: 56974-61-9
Supplier: Enzo Life Sciences BMLPI1440050
A competitive, reversible inhibitor of serine proteases tryptase (Ki=3.4nM), thrombin (Ki=500nM), uPA (Ki=1.3µM), plasmin (Ki=1.5µM), trypsin (Ki=2µM), factor Xa (Ki=2µM). At higher concentrations it inhibits kallikrein-1 (Ki=2.6mM), C1 esterase (IC50=520µM), as well as phospholipase A2 (IC50=500µM), rat iNOS (Ki=5mM) and cNOS (Ki=150µM), and swine kidney copper amine oxidase (Ki=27µM), but not chymotrypsin. Has been used in tissue culture and in vivo. In clinical use for pancreatitis. Purity: ≥98% (HPLC). Solubility: Soluble in water (25 mg/ml). Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.