Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galactitol 98.0+%, TCI America™
SDP

Catalog No. p-7130095 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
250 g

Chemical Identifiers

CAS 608-66-2
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.172
MDL Number MFCD00064288
InChI Key FBPFZTCFMRRESA-GUCUJZIJSA-N
Synonym dulcitol, galactitol, dulcite, d-galactitol, dulcose, euonymit, melampyrin, melampyrit, d-dulcitol, 2r,3s,4r,5s-hexane-1,2,3,4,5,6-hexaol
PubChem CID 11850
ChEBI CHEBI:16813
IUPAC Name (2R,3S,4R,5S)-hexane-1,2,3,4,5,6-hexol
SMILES C(C(C(C(C(CO)O)O)O)O)O

Specifications

Melting Point 189°C
Color White
Boiling Point 280°C
Quantity 25 g
Formula Weight 182.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Galactitol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.