Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallic Acid, MP Biomedicals

Catalog No. p-3593012 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 g
250 g

Chemical Identifiers

CAS 149-91-7
Molecular Formula C7H6O5
Molecular Weight (g/mol) 170.12
MDL Number MFCD00002510
InChI Key LNTHITQWFMADLM-UHFFFAOYSA-N
Synonym gallic acid, gallate, benzoic acid, 3,4,5-trihydroxy, gallic acid, tech., galop, pyrogallol-5-carboxylic acid, kyselina gallova, 3,4,5-trihydroxybenzoate, ccris 5523, kyselina gallova czech
PubChem CID 370
ChEBI CHEBI:30778
IUPAC Name 3,4,5-trihydroxybenzoic acid
SMILES OC(=O)C1=CC(O)=C(O)C(O)=C1

Specifications

Color White
Formula Weight 170.1
Density 1.694g/mL (Lit.)
Quantity 100 g
Physical Form Solid
Chemical Name or Material Gallic Acid
EINECSNumber 205-749-9
Recommended Storage Store desiccated. Protect from light. Store at room temperature (15° to 30°C).
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.