Learn More
TARGETMOL CHEMICALS INC GALLOPAMIL HYDROCHLORIDE 25MG
Also available in 1 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Gallopamil hydrochloride (Methoxyverapamil hydrochloride) is an antagonist of phenylalkylamine calcium. Gallopamil hydrochloride can be used in antiarrhythmic and vasodilator studies. purity: 99%