Learn More
Enzo Life Sciences Gambogic acid (5mg). CAS: 2752-65-0
SDP
Supplier:  Enzo Life Sciences BMLAP3050005
Encompass_Preferred
Gambogic Acid is a natural product isolated from Garcinia Hurburyi. It induces apoptosis in several tumor cell lines including T47D breast cancer cells and MGC-803 human gastric carcinoma cells but not in normal cells. Apoptosis induction proceeds via upregulation of bax and inhibition of bcl-2 expression.The target of gambogic acid has been identified as the transferrin receptor. Purity: ≥95% (TLC). Solubility: Soluble in DMSO (25 mg/ml) or ethanol (25 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0314
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.