Learn More
Medchemexpress LLC HY-13637 5g , Ganciclovir BW 759;2'-Nor-2'-deoxyguanosine CAS:82410-32-0 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY136375G
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-13637 5g Ganciclovir BW 759;2'-Nor-2'-deoxyguanosine CAS:82410-32-0 Formula:C9H13N5O4 IC50: 5.2 μM (FHV-1) Purity:98% Ganciclovir is a potent inhibitor of viruses of the herpes family, including cytomegalovirus ( CMV ), with an IC 50 of 5.2 μM for feline herpesvirus type-1 (FHV-1). Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2228
missing translation for 'QuantityLeft'