Learn More
MilliporeSigma™ Supelco™ Gas Sampling Bulbs
For trapping and transporting gas samples for analysis
Supplier: MilliporeSigma™ Supelco™ 22148U
Description
- Use the plug-type septum to easily remove a gas sample for analysis
- Oven annealed to resist damage during transportation and on-site use
- Each bulb includes one half-hole, cylindrical Thermogreen™ septum
- Available with PTFE or high vacuum glass stopcocks
Specifications
Glass Stopcock | |
1 Each | |
500 mL |
Trapping and Transporting Gas Samples for Analysis | |
Gas Sampling Bulb |