Learn More
Cole-Parmer Wika 232.54 2.5 Industrial Stainless Steel Gauge, 0 to 15 psi
Supplier: Cole-Parmer 6843272
These gauges feature 316 stainless steel wetted parts and 304 stainless steel case and bayonet ring, an adjustable pointer, a laminated safety glass window, and can be liquid-filled in the field. WIKA stainless steel gauges are ideal for process, chemical, and petrochemical applications, oil exploration and production, power generation, pollution control equipment, and any other application requiring fine quality, long lasting pressure measurement instrumentation.
- Center Back Mount Connection Available
- 1/4" NPT Connection Size
- 4" Dial Size Available
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.