Learn More
Gold Biotechnology Inc GB5-alpha Electrocompetent E. coli Cells 5x100 μl
SDP
Supplier:  Gold Biotechnology Inc CC2035X100C
Encompass_Preferred
GoldBio’s GB5-alpha Electrocompetent E. coli are high efficiency cells, suitable for a wide variety of applications such as cloning and sub-cloning. Plasmid quality and yields are both enhanced in our GB5-alpha cells due to mutations in both endA1 and recA1.
Catalog No. NC2942857
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.