Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GBR 12783 dihydrochloride, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
A very potent and selective inhibitor of dopamine uptake (IC50 for inhibition of [3H]-dopamine uptake in rat striatal synaptosomes is 1.8 nM).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 485.5 |
Percent Purity | >99% |
Chemical Name or Material | GBR 12783 dihydrochloride |
Safety and Handling
Recommended Storage | Desiccate at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction