Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDC 0879, Tocris Bioscience™

Catalog No. p-4816999 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent B-Raf inhibitor

Potent and selective B-Raf inhibitor (IC50 = 0.13 nM against purified B-Raf V600E). Activity reduces phospho-ERK (pERK) levels (IC50 = 63 nM in the Malme-3M cell line). Inhibits the Raf/MEK/ERK signaling pathway in V600E B-Raf mutant cell lines. Does not activate apoptotic pathways in A375 or Colo205 cell lines. Orally bioavailable.

Chemical Identifiers

CAS 905281-76-7
Molecular Formula C19H18N4O2
Molecular Weight (g/mol) 334.379
InChI Key DEZZLWQELQORIU-UHFFFAOYSA-N
Synonym e-5-1-2-hydroxyethyl-3-pyridin-4-yl-1h-pyrazol-4-yl-2,3-dihydroinden-1-one oxime, 2-4-1-hydroxyimino-2,3-dihydroinden-5-yl-3-pyridin-4-yl pyrazol-1-yl ethanol, 5-1-2-hydroxyethyl-3-pyridin-4-yl-1h-pyrazol-4-yl-2,3-dihydroinden-1-one oxime
PubChem CID 57519545
IUPAC Name 2-[4-(1-hydroxyimino-2,3-dihydroinden-5-yl)-3-pyridin-4-ylpyrazol-1-yl]ethanol
SMILES C1CC(=NO)C2=C1C=C(C=C2)C3=CN(N=C3C4=CC=NC=C4)CCO

Specifications

Quantity 10 mg
Formula Weight 334.37
Percent Purity >98%
Chemical Name or Material GDC 0879
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.