Learn More
Electron Microscopy Sciences Gel Drying Frames 24 x 24cm Gel Frame
Supplier:  Electron Microscopy Sciences 6851024
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Gel Drying Frames allow researchers to quickly and easily dry gels between two sheets of cellophane without the use of expensive heat/vacuum gel dryers. 24 x 24cm frames allow for drying of larger format gels. Kit: 2 open frames, 2 solid back plates, 12 side clips, 50 sheets pre-cut cellophane.
Catalog No. 50-345-29
missing translation for 'QuantityLeft'