Learn More
Medchemexpress LLC HY-B0003 1g , Gemcitabine (Hydrochloride) LY 188011 hydrochloride CAS:122111-03-9 Purity:98%
Medchemexpress, HY-B0003 1g Gemcitabine (Hydrochloride) LY 188011 hydrochloride CAS:122111-03-9 Formula:C9H12ClF2N3O4 DNA synthesis Purity:98% Gemcitabine hydrochloride is a DNA synthesis inhibitor with IC 50 of 37.6, 42.9, 92.7, 89.3 and 131.4 nM in BxPC-3, Mia Paca-2, PANC-1, PL-45 and AsPC-1 cells, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.