Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Gemfibrozil, 98%

Catalog No. AC461790250
Click to view available options
Quantity:

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS

25812-30-0

Molecular Formula

C15H22O3

Molecular Weight (g/mol)

250.34

MDL Number

MFCD00079335

InChI Key

HEMJJKBWTPKOJG-UHFFFAOYSA-N

Synonym

gemfibrozil, lopid, 5-2,5-dimethylphenoxy-2,2-dimethylpentanoic acid, jezil, decrelip, lipur, bolutol, apo-gemfibrozil, gemfibromax, cholespid

PubChem CID

3463

ChEBI

CHEBI:5296

IUPAC Name

5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoic acid

SMILES

CC1=CC=C(C)C(OCCCC(C)(C)C(O)=O)=C1

Specifications

Melting Point

60.0°C to 61.0°C

Quantity

25 g

Assay Percent Range

97.5% min. (HPLC)

Formula Weight

250.33

Percent Purity

98%

Chemical Name or Material

Gemfibrozil

Hazard Category

GHS Signal Word: Warning

Hazard Statement

GHS H Statement
Harmful if swallowed.

Precautionary Statement

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.

EINECSNumber

247-280-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.