Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GeneArt™ Gibson Assembly HiFi Cloning Kit, chemically competent cells
SDP

Catalog No. A46624EA
Click to view available options
:

The GeneArt Gibson Assembly HiFi Cloning Kit enables DNA assembly and cloning via a technique that allows multiple overlapping DNA fragments to be seamlessly linked in a one-step, 15-60 minute isothermal reaction.

The inherent flexibility of this approach is suitable for small and large DNA constructs and includes both single and multiple inserts. The resulting products can be used for a variety of downstream applications including transformation, PCR and rolling circle amplification (RCA). The GeneArt Gibson Assembly HiFi Cloning Kit, chemically competent cells, is a complete kit that includes master mix, positive control, water, and One Shot TOP10 chemically competent E. coli.

Features of the GeneArt Gibson Assembly HiFi Cloning Kit include:
Simple—seamlessly assemble and clone up to six DNA fragments in a single reaction
Efficient—high fidelity provides more correct clones than other methods
Flexible—design guidelines allow assembly into any vector of your choice
Convenient—available as master mix kits and cloning kits complete with chemical or electrocompetent cells
Trusted—over 4000 citations in the scientific literature highlighting great success

Seamless assembly of multiple fragments
GeneArt Gibson Assembly HiFi cloning is a simple, one-step process whereby up to six fragments are combined in a proprietary enzymatic mix in order to assemble DNA fragments with shared terminal end homology without leaving any extra sequences or scars behind (seamless). After a 15–60 minute incubation, a portion of the assembly reaction is then transformed into chemically competent or electrocompetent cells, yielding clones that are ready for analysis the next day. The required 20- to 40-base pair end homology is designed into the de novo fragment for synthesis or can be easily engineered by PCR amplification with custom DNA oligos. This special enzyme mix creates a seamless and covalently bound DNA construct providing high efficiency.

Robust method for maximum efficiency
Due to the covalently bound final product, the GeneArt Gibson Assembly HiFi method allows the utilization of chemically competent cells or electrocompetent cells for the highest transformation efficiency, improving the success of finding the right clone, particularly for more challenging constructs. There is no need for restriction enzymes, ligation, or recombination sites, and the method provides a perfectly seamless construct without unwanted extra bases.

Provides great versatility
The GeneArt Gibson Assembly HiFi method provides versatility, can streamline many techniques through the rapid combination, addition, deletion, or exchange of DNA segments, and eliminates the need to subclone, saving time and effort in the cloning workflow.

Specifications

Bacterial or Yeast Strain

TOP10

Cell Type

Chemically Competent E. coli

Cloning Method

Seamless cloning

Content And Storage

Includes master mix, positive control, water, and One Shot TOP10 chemically competent E. coli.

Control Type

Positive Control

Format

Kit

For Use With (Application)

Seamless Cloning & Genetic Assembly

No. of Reactions

10 Reactions

Number of Fragments

Up to 6 fragments

Product Line

GENEART™

Product Type

Cloning Kit

Shipping Condition

Dry Ice

Workflow Step

DNA Assembly

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.