Learn More
Electron Microscopy Sciences Geological Thin Section Holder Hitachi M4
Supplier: Electron Microscopy Sciences 7594115H
This thin section holder is made from aluminum with copper clips and brass screws. The unit measures 55x30x8 mm and holds a standard geological thin section or slide measuring 47 x 27 mm. The holder comes complete with 2 clips to insure nothing moves. The holder has convenient tweezer inserts which allow for easy loading.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.