Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GF 109203X, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
1 mg
10 mg
Description
Very potent and selective inhibitor of protein kinase C, selective for the α and β1 isoforms (IC50 values are 0.0084, 0.0180, 0.210, 0.132, and 5.8 μM for α, β1, δ, ε and ζ isoforms respectively). Selective over MLCK, PKG and PKA (IC50 values are 0.6, 4.6, and 33 μM respectively). Potent antagonist at the 5-HT3 receptor (Ki = 29.5 nM). Anti-inflammatory in vivo.
Specifications
Specifications
Quantity | 1 mg |
Formula Weight | 412.49 |
Percent Purity | >98% |
Chemical Name or Material | GF 109203X |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction