Learn More
Medchemexpress LLC HY-B0355 10mg Medchemexpress, Ginkgolide A CAS:15291-75-5 Purity:>98%
Supplier: Medchemexpress LLC HYB035510MG
Medchemexpress, HY-B0355 10mg Ginkgolide A CAS:15291-75-5 Ginkgolide A (BN-52020) is an extract from in Ginkgo biloba and a g-aminobutyric acid (GABA) antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.