Learn More
Medchemexpress LLC HY-N0393 10mg Medchemexpress, Glabridin CAS:59870-68-7 Purity:>98%
Medchemexpress, HY-N0393 10mg Glabridin CAS:59870-68-7 Glabridin is a natural isoflavan from Glycyrrhiza glabra, binds to and activates PPARγ , with an EC 50 of 6115 nM. Glabridin exhibits antioxidant, anti-bacterial, anti-nephritic, anti-diabetic, anti-fungal, antitumor, anti-inflammatory, antiosteoporotic, cardiovascular protective, neuroprotective and radical scavenging activities [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.