Learn More
U.S. Pharmacopeia Glacial Acetic Acid, 64-19-7, MFCD00036152, 1.5 mL/ampule; 3 ampules
Supplier:  U.S. Pharmacopeia 1005706
Encompass
Molecular formula C2-H4-O2, Molecular weight 60.05 g/mol, Melting Point 61.88 °F (16.6 °C), Boiling Point 244.22 °F (117.9 °C), Synonyms: Methanecarboxylic acid, Ethanoic acid
Catalog No. NC9871853
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.