Learn More
Eisco™ Glass Burette with Needle Valve Stopcock, Sure Trace Certification
Class A, DIN 385 Compliant, Borosilicate Glass with PTFE Needle Valve Stopcock, with graduations.
Supplier:  Eisco™ CH0238DST
 View more versions of this product
Catalog No. 12-183-070
Description
Description
  • Accurate as per class A, DIN ISO 385 Compliant
  • Includes an attached PTFE needle valve rotaflow stopcock
  • Burette is made of precisely bore high quality, heavy duty Borosilicate 3.3 glass
  • Features chemical resistant blue printing - All contents are included in a single package
Specifications
Specifications
100 mL
Borosilicate Glass (3.3)
Class A
86.36 cm
Buret
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.