Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucotropaeolin Potassium Salt 95.0+%, TCI America™
SDP

Catalog No. G039710MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
10 mg

Chemical Identifiers

CAS 5115-71-9
Molecular Formula C14H18KNO9S2
Molecular Weight (g/mol) 447.514
MDL Number MFCD00153006
InChI Key UYCWNAZWHVREMO-GYVLLFFHSA-M
PubChem CID 70702337
IUPAC Name potassium;[(E)-[2-phenyl-1-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]sulfanylethylidene]amino] sulfate
SMILES C1=CC=C(C=C1)CC(=NOS(=O)(=O)[O-])SC2C(C(C(C(O2)CO)O)O)O.[K+]

Specifications

Quantity 10 mg
Formula Weight 447.51
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Glucotropaeolin Potassium Salt
TSCA No
Recommended Storage Freezer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.