Learn More
Medchemexpress LLC HY-100111 100mg Medchemexpress, GLX351322 CAS:835598-94-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100111100MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-100111 100mg GLX351322 CAS:835598-94-2 GLX351322 is an inhibitor of NADPH oxidase 4 ( Nox4 ), and inhibits hydrogen peroxide production from NOX4-overexpressing cells with an IC 50 of 5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1717
missing translation for 'QuantityLeft'