Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Glycerin
SCI_ED

Catalog No. p-4416552 Shop All Fisherbrand Products
Click to view available options
Quantity:
50 mL
500 mL
1 L
4 L
Packaging:
ACS Grade - Liquid
Lab Grade - Liquid

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Density 1.2610g/°Cm3 at 20°C
Assay Percent Range 99 to 100% w/v
Packaging ACS Grade - Liquid
Linear Formula C3H5(OH)3
Quantity 500 mL
Formula Weight 92.1
Grade ACS
Chemical Name or Material Glycerin
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.