Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerol Dimethacrylate (mixture of 1,2- and 1,3-form) (stabilized with MEHQ) 90.0+%, TCI America™
SDP

Catalog No. p-7119851 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 1830-78-0
Molecular Formula C11H16O5
Molecular Weight (g/mol) 228.24
MDL Number MFCD00066299
InChI Key OQHMGFSAURFQAF-UHFFFAOYSA-N
Synonym Bis(methacryloyloxy)propanol
PubChem CID 74594
IUPAC Name 2-hydroxy-3-[(2-methylprop-2-enoyl)oxy]propyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(O)COC(=O)C(C)=C

Specifications

Color Yellow
Boiling Point 104°C
Quantity 25 g
Formula Weight 228.24
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Glycerol Dimethacrylate (mixture of 1,2- and 1,3-form) (stabilized with MEHQ)
EINECSNumber (2)-1063&(2)-0958
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.