Learn More
Cell Biolabs Inc GLYCOGEN ASSAY KT FLUOR 100 RX
Supplier: Cell Biolabs Inc MET5023
The Glycogen Assay Kit (Fluorometric) measures glycogen in serum, plasma, urine, lysates, and cell culture supernatants. The hydrogen peroxide released during the assay reacts specifically with the kit’s Fluorometric Probe.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.