Learn More
Selleck Chemical LLC GNF-2 5mg 778270-11-4
Supplier:  Selleck Chemical LLC S28995MG
Encompass_Preferred
GNF-2 is a highly selective non-ATP competitive inhibitor of Bcr-Abl, shows no activity to Flt3-ITD, Tel-PDGFR, TPR-MET and Tel-JAK1 transformed tumor cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8151
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.