Learn More
Electron Microscopy Sciences Gold Chloride 0.2% Aqueous 250 ML
Supplier:  Electron Microscopy Sciences 2639209
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Used in Wilder Modification of Bielschowsky's Method for staining of Collagen and reticulum. Reticulum Fibers (If beading is observed and/or a dark background, replace the ammonium hydroxide silver. References: Wilder, H.C., Amer. J. Clin. Path.: 11:817, 1935, Mallory, F.B.: Pathological Techniques, Phil., W.B. Saunders & Co. c. 1938, or AFIP Manual of Hitological Staining Techniques, NY, McGraw-Hill Publ., c. 1968/3rd ed, p. 92
Catalog No. 50-317-97