Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Gold Chloride, Hydrate 1 GR

Supplier:  Electron Microscopy Sciences 16584

Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred

CAS #16961-25-4 H(AuCl₄)·H₂O CAS F.W. 357.79
Assay: 99.9985%

A highly purified gold compound for the preparation of Colloidal Gold sols. Preweighed to facilitate your work. (Preparation instructions for Gold Sols included).

Catalog No. 50-263-21


missing translation for 'QuantityLeft'
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.