Learn More
Ted Pella Inc GOLD COLLOID (SOLS) 20ML 10NM
SDP
Supplier:  Ted Pella Inc 1570320
Encompass
Gold Colloids, 10nm, 5.7 x 1012, 20mL, Room Temperature
Catalog No. NC9851179
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.