Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold Germanium slug, 3.175mm (0.125 in.) dia. X≈3.175mm (0.125 in.) length, Premion™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7058668
Click to view available options
Quantity:
25 Pc.
5 Pc.

Gold Germanium slug is used in scientific research and development activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Gold Germanium slug
Name Note 3.175mm (0.125 in.) dia. x≈3.175mm (0.125 in.) length
Avg. Mol. Wt. or Mol. Wt. Range Au:Ge; 88:12 wt%
Density 0.37 g/cm3
Assay Percent Notes (metals basis)
MDL Number MFCD02091706
Quantity 5 Pc.
Solubility Information Insoluble in water.
Percent Purity 99.998%
Grade Premion™
Odor Odorless
Weight ≈0.4g/slug
Form Slug
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.