Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roylco™ Goo Spreaders
SCI_ED

Catalog No. S13212
Click to view available options
:
Goo Spreaders; Material: Plastic

Goo spreaders are ideal tools for tiny hands to spread glue and mix paint.

  • Takes the mess out of craft time
  • Spreads glue, paint and more
  • A handy tool for every classroom
  • Useful in art projects
  • Assorted colors
  • Comes with idea guide

Specifications

Grade 1+
Format Kit
Quantity 10 Tools/Pk.
Material Plastic
Languages English, Spanish, French, German
Subject My Senses
Length (English) 4 in.
Length (Metric) 13 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.