Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GR 127935 hydrochloride, Tocris Bioscience™

Catalog No. p-4783490 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective 5-HT1B and 5-HT1D antagonist

Potent and selective 5-HT1B/1D receptor antagonist (pKi values are 8.5 for both guinea pig 5-HT1D and rat 5-HT1B receptors). Displays > 100-fold selectivity over 5HT1A, 5-HT2A, 5-HT2C receptors and other receptor types. Centrally active following oral administration.

Chemical Identifiers

CAS 148642-42-6
Molecular Formula C29H32ClN5O3
Molecular Weight (g/mol) 534.057
InChI Key SRVVUYIJVBLEJI-UHFFFAOYSA-N
PubChem CID 11497466
ChEBI CHEBI:64111
IUPAC Name N-[4-methoxy-3-(4-methylpiperazin-1-yl)phenyl]-4-[2-methyl-4-(5-methyl-1,2,4-oxadiazol-3-yl)phenyl]benzamide;hydrochloride
SMILES CC1=C(C=CC(=C1)C2=NOC(=N2)C)C3=CC=C(C=C3)C(=O)NC4=CC(=C(C=C4)OC)N5CCN(CC5)C.Cl

Specifications

Quantity 10 mg
Synonym gr 127935 hydrochloride, gr127935 hydrochloride, gr 127935 hydrochloride hydrate, n-4-methoxy-3-4-methyl-1-piperazinyl phenyl-2'-methyl-4'-5-methyl-1,2,4-oxadiazol-3-yl-1,1'-biphenyl-4-carboxamide hydrochloride, n-4-methoxy-3-4-methylpiperazin-1-yl phenyl-2'-methyl-4'-5-methyl-1,2,4-oxadiazol-3-yl biphenyl-4-carboxamide hydrochloride, 4-2-methoxy-5-2'-methyl-4'-5-methyl-1,2,4-oxadiazol-3-yl biphenyl-4-yl carbonyl amino phenyl-1-methylpiperazin-1-ium chloride
Formula Weight 534.06
Percent Purity >99%
Chemical Name or Material GR 127935 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.