Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GR 55562 dihydrochloride, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
A selective competitive 5-HT1B (5-HT1Dβ) silent antagonist with pKB values of 7.3 and 6.3 for human cloned 5-HT1B and 5-HT1D receptors respectively and only weak binding at a number of other 5-HT subtypes.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 448.39 |
Percent Purity | >98% |
Chemical Name or Material | GR 55562 dihydrochloride |
Safety and Handling
Recommended Storage | Desiccate at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction