Learn More
SP Hotpack™ Graduated Cylinder Basket Insert
Accessories designed for use with Hotpack™ Undercounter, Freestanding and Vertical Stacking Glassware washers safeguard flasks, beakers, test tubes, bottles, petri dishes, and more.
Supplier: SP Hotpack™ 437165
Description
- Up to 32 small graduated cylinders, 1/2 of rack, top or bottom
- Accessories maximize space, improve efficiency, and protect specialty glassware
- Use in standard open racks
- Bottom mesh inserts prevent small items from falling through
- Made of stainless steel for long life and durability
- Booster Pump for use with low pressure DI water supply below 18 psi
Specifications
Holds up to 32 graduated cylinders | |
Stainless Steel | |
Glassware Washer Accessory |